Learn More
Andwin Scientific DRUMMOND PIPET AID XP
SDP
Catalog No. NC9377742
Encompass_Preferred
The Drummond Pipet-Aid XP controller is made to easily and efficiently fill and empty pipettes. The speed of the pipet filler can be easily and conveniently modified to fast, medium, or slow. With a weight of 7.5 ounces, this is also much lighter than the previous models. This product is sold individually with a charger and stand.
Catalog No. NC9377742 Supplier Andwin Scientific Supplier No. P30731
The Drummond Pipet-Aid XP controller is made to easily and efficiently fill and empty pipettes. The speed of the pipet filler can be easily and conveniently modified to fast, medium, or slow. With a weight of 7.5 ounces, this is also much lighter than the previous models. This product is sold individually with a charger and stand.