Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drummond™ Portable Pipet-Aid™ XL Pipet Controller Accessory, EZ Grip Nosepieces

Catalog No. 2117117
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-171-17 Blue
21-171-19 Orange
21-171-18 Green
21-171-20 Red
21-171-21 Purple
21-171-22 Yellow
21-171-3 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 21-171-17 Supplier Drummond™ Supplier No. 4000079B
Add to Cart
Add to Cart

Replacement Part for Pipet-Aid™ Filler/Dispensers

  • Choice of colors: Orange, blue, red, violet, yellow, green
  • Autoclavable

Specifications

Autoclavable Not Autoclavable
Color Blue
For Use With (Equipment) Pipet-Aid™ XP, XL, and Hood Mate™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.