Learn More
Drummond™ Nanoject II Auto-Nanoliter Injector
Catalog No. 13681455
Change view
Click to view available options
Nanoject II Auto-Nanoliter Injector
Catalog No.
13-681-455 Nanoject II Auto-Nanoliter Injector
1 options
Catalog No. 13-681-455 Supplier Drummond™ Supplier No. 3000204
Includes
Universal adapter 3-000-024-A
Specifically designed to perform ultra-delicate nanoliter injection
  • Selectable operating speeds with high and low settings: Fill speed (46 and 23nL/s), injection speed (46 and 23nL/s), and empty speed (230 and 92nL/s)
  • Selectable injection volumes range from 2.3nL to 69.0nL in 16 increments
  • Automated microprocessor controlled microinjection pipet delivers precise nanoliter volumes
  • Remote control and nonrotation plunger eliminates vibration, minimizing possible cell rupture
  • Supplied borosilicate glass capillaries are capable of preparing 400 injection tips, standard softening point 780°C
  • Able to be mounted on most micromanipulators
Specifications
Description Controller: Automated microprocessor; Speed: 46 (Fast) and 23 (slow) for fill and injection, 230 (fast) and 92 (slow) for empty
Electrical Requirements 110 V
For Use With (Application) Specifically designed to perform ultra-delicate nanoliter injection procedures into oocytes, embryos and tissues
Graduations 16 mL
Includes Universal adapter, Borosilicate Glass capillaries, remote control and nonrotation plunger
Type Nanoject II™ Auto-nanoliter injector
For Use With (Equipment) Micromanipulator