Learn More
Drummond™ Dispensing Manifold
GSA_VA
Catalog No. 2116911B
Change view
Click to view available options
Angle:
No
Yes
Number of Channels:
8
12
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Angle Number of Channels
21-169-11B Yes 12
21-169-11A No 12
21-169-10D No 8
21-169-10E Yes 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-169-11B Supplier Drummond™ Supplier No. 3000099
Add to Cart
Add to Cart
Eight-tip manifolds for use with the 21-169-10A Digital Microdispenser or any pipetter that accepts standard Luer-Lok™ connector
  • Stainless steel and PTFE construction
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Angle Yes
Description 9mm (center to center); 11mm pin length
Material Stainless Steel, PTFE
Number of Channels 12
For Use With (Equipment) Drummond™ micropipetter
Includes Cleaning wire, Luer connector and end caps