Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drummond™ Wiretrol™ I Calibrated Micropipets

Catalog No. 21175C
Change view
Click to view available options
Volume (Metric):
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-175C 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-175C Supplier Drummond™ Supplier No. 50001020

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Handy dispenser cap, handle, stainless-steel plunger

Easy-to-read volume line won't wash off

  • Deliver within 1%
  • One-handed operation
  • Accepts both high- and low-viscosity liquids including corrosives
  • Color coded
  • Micropipet 21-175D, -175E and -175F have PTFE-fluorocarbon–resin tips on plunger

Compliance

Calibrated To Contain

Specifications

Calibration To Contain
Material Glass
For Use With (Application) For a wide variety of applications, Wiretrol™ will accept both high and low viscosity liquids including corrosives
Includes Handy dispenser cap, handle, Stainless Steel plunger
Color Black
Volume (Metric) 20 μL
Type Calibrated Micropipet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.