Learn More
Cole-Parmer Drawer Freezer Bin Rack Medium, 22.01 x 9.92 x 5.63" (L x H x W)
Catalog No. NC1434701 Shop All Cole-Parmer Products
Encompass_Preferred
Organize and Optimize your Laboratory FreezerPolarSafe Upright Freezer Bin Racks are are designed to store miscellaneous bottles, tubes, vials, bags or boxes. Racks sides are made from wire mesh to allow for easy viewing of contents, the open format works well with -20C freezers. Racks are available in both an upright (including lid) and drawer style. The drawer style racks are fully framed and work like standard drawer racks. All Upright Freezer Bin racks are constructed of high quality corrosion resistant stainless steel.
Catalog No. NC1434701 Supplier Cole-Parmer Supplier No. 0439796
Organize and Optimize your Laboratory FreezerPolarSafe Upright Freezer Bin Racks are are designed to store miscellaneous bottles, tubes, vials, bags or boxes. Racks sides are made from wire mesh to allow for easy viewing of contents, the open format works well with -20C freezers. Racks are available in both an upright (including lid) and drawer style. The drawer style racks are fully framed and work like standard drawer racks. All Upright Freezer Bin racks are constructed of high quality corrosion resistant stainless steel.