Learn More
Brookfield Engineer Labs DRY AIR GENERATOR KIT
Catalog No. NC3467093
Encompass
  • Provides a self-regulating source of dry compressed air.
  • Streamlines setup and operation by eliminating the need for an external regulator.
  • Ideal for portable applications, ensuring consistent performance.
  • Supports sensitive analytical equipment.
  • Output pressure of up to 20 psi.
  • Input voltage of 115 VAC.
  • Warranty period of 1 year.
Catalog No. NC3467093 Supplier Brookfield Engineer Labs Supplier No. Y9900143
  • Provides a self-regulating source of dry compressed air.
  • Streamlines setup and operation by eliminating the need for an external regulator.
  • Ideal for portable applications, ensuring consistent performance.
  • Supports sensitive analytical equipment.
  • Output pressure of up to 20 psi.
  • Input voltage of 115 VAC.
  • Warranty period of 1 year.