Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dry-Bags™ Pump Tubing

Catalog No. 21377818 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With:
Dry-Bags
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
21-377-818 Dry-Bags
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-377-818 Supplier Thermo Scientific™ Supplier No. DB1750A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use this platinum-cured silicone tubing with Thermo Scientific™ peristaltic pump units to deliver water from its source to Thermo Scientific™ Dry-Bags™.

Effectively deliver water from its source to Thermo Scientific™ Dry-Bags™ using platinum-cured silicone Thermo Scientific Dry-Bags Pump Tubing.

Use with Thermo Scientific Peristaltic Pump units DB2000A and DB3000A. Use Dry-Bags Tubing Connector BM9901A to attach Dry-Bags Pump Tubing to the Dry-Bags filling tubing.

  • Excellent biocompatibility, autoclavable and durable
  • No leachable additives or plasticizers
  • Phthalate and latex-free
  • Odorless and nontoxic
  • Fungus-resistant
  • Extremely durable over a wide temperature range -50 to 230°C (-58 to 446°F)

Clean with hot water/soap solution; use a non-oily soap such as Ivory™, not synthetic detergent or oil-based soap as they may be absorbed by the tubing and into the fluid. Rinse well with distilled water.

Contact us to arrange a demonstration of the Peristaltic Pump and its accessories.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP Class V Extractables; exceeds Class VI Implant; FDA 21 CFR 177.2600; Exceeds 3A Sanitary cGMPs (FDA 21 CFR 210 and 211). European Pharmacopoeia (EP)
Description Dry-Bags Pump Tubing
Autoclavable Autoclavable
Material Thermal Set Rubber (Platinum-cured Silicone)
For Use With Dry-Bags
Includes Includes pump tubing only
Color Translucent Clear to Light Amber
Max. Operating Temperature (English) 446°F
Max. Operating Temperature (Metric) 230°C
Product Type Pump Tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.