Learn More
Thermco Products Inc Special application Non-Hg, 0 deg./110 deg.C, 0.5C div, 35mm imm, 225mm
Catalog No. NC0351666
Encompass_Preferred
Precision spirit filled thermometer PTFE coated for safety to contain the glass and liquid if broken. The ACC647S thermometer was designed to be used in block heater (dry baths) with it's 35mm immersion. Accurate to N.I.S.T. tolerances. Eligible for N.I.S.T. certification. Also used in shallow water baths
Catalog No. NC0351666 Supplier Thermco Products Inc Supplier No. ACC647SSC
Precision spirit filled thermometer PTFE coated for safety to contain the glass and liquid if broken. The ACC647S thermometer was designed to be used in block heater (dry baths) with it's 35mm immersion. Accurate to N.I.S.T. tolerances. Eligible for N.I.S.T. certification. Also used in shallow water baths