Learn More
Cole-Parmer Metallic Bath Beads for Dry Baths; 4-Liters
Supplier:  Cole-Parmer 0627451
Encompass_Preferred
Hold containers in place at any angle without any float-away or ruined samples. High thermal conductivity 180C. Versatile enough for freezers, ovens and incubators. Smooth, rounded surfaces measure 5 to 8 mm diameter and 1 to 2 mm high. Beads are moisture and gas impermeable and are easy to keep clean without harmful germicides. These versatile beads can also be used in a container for freezer, ovens and incubators.
Catalog No. NC0632526