Learn More
Electron Microscopy Sciences EMS MyBlock™ Dry Bath I Single Position universal block 230 V
SDP
Catalog No. 501927874
Encompass_Preferred
The EMS MyBlock series has several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of temperature.
MyBlock's transparent lid promotes better temperature uniformity and minimizes stratification in the block by maintaining a warm air jacket layer while reducing energy consumption by minimizing heat loss. The hinged lid can be flipped open or easily removed (lifted off) to accommodate taller tubes.
Universal blocks accept tubes from 0.5 to 2.0 ml and, with just a quick flip to the other side, accomodate 0.2 ml tubes, PCR strips or in the dual block model only a PCR plate. Other blocks are available for tubes up to 50 ml. With 1 Quick-Flip universal block.
Catalog No. 50-192-7874 Supplier Electron Microscopy Sciences Supplier No. 8702606
The EMS MyBlock series has several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of temperature.
MyBlock's transparent lid promotes better temperature uniformity and minimizes stratification in the block by maintaining a warm air jacket layer while reducing energy consumption by minimizing heat loss. The hinged lid can be flipped open or easily removed (lifted off) to accommodate taller tubes.
Universal blocks accept tubes from 0.5 to 2.0 ml and, with just a quick flip to the other side, accomodate 0.2 ml tubes, PCR strips or in the dual block model only a PCR plate. Other blocks are available for tubes up to 50 ml. With 1 Quick-Flip universal block.