Learn More
Cole-Parmer Techne Tecal 425F Portable/Field Dry Block Calibrator, 425°C; 120 VAC
Catalog No. 501961444 Shop All Cole-Parmer Products
Encompass_Preferred
These lightweight, portable dry block temperature calibrators are ideal for use in the field. They can be used with a wide range of inserts (not included) with different dimension wells to match numerous probe sizes. Safety features include an automatic cooling fan and overtemperature cutout which turns power off to the heaters if a fault develops or the unit exceeds maximum operating temperature. Temperature can be displayed in Celsius or Fahrenheit.
Catalog No. 50-196-1444 Supplier Cole-Parmer Supplier No. 1610215
These lightweight, portable dry block temperature calibrators are ideal for use in the field. They can be used with a wide range of inserts (not included) with different dimension wells to match numerous probe sizes. Safety features include an automatic cooling fan and overtemperature cutout which turns power off to the heaters if a fault develops or the unit exceeds maximum operating temperature. Temperature can be displayed in Celsius or Fahrenheit.