Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Dry Block Heaters
GSA_VA

Catalog No. 021061018 Shop All OHAUS Products
To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.
Catalog No. 02-106-1018 Supplier OHAUS™ Supplier No. 30392080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.

OHAUS™ Dry Block Heaters are designed for applications that require repeatable results and superior temperature stability

  • Dry block heaters require either one or two modular blocks, sold separately
  • Ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 Blocks
Temperature Range (Metric) Ambient +5° to 120°C
Type Block
For Use With (Application) Ideal for incubation and activation of cultures, Enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedure
Frame Material Steel
Heat Up Time 50 min.
Includes Detachable, 3-wire cord and plug (92 in.)
Temperature Uniformity ±0.1°C
Length (English) 15.4 in.
Length (Metric) 39.1 cm
Height (English) 3.5 in.
Height (Metric) 8.9 cm
Weight (English) 6.4 lb.
Weight (Metric) 2.9 kg
Electrical Requirements 120 V, 1.7 A, 210 W
Amperage 1.7 A
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.