Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Dry Block Heaters
SCI_ED

Catalog No. S38783 Shop All OHAUS Products
Change view
Click to view available options
Electrical Requirements:
120 V, 0.9 A, 110 W
120 V, 1.7 A, 210 W
Holds:
1 Block
2 Blocks
Temperature Range (Metric):
Ambient +5° to 120°C
Ambient +5°C to 150°C
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrical Requirements Holds Temperature Range (Metric)
S38783 120 V, 0.9 A, 110 W 1 Block Ambient +5° to 120°C
S38780 120 V, 1.7 A, 210 W 2 Blocks Ambient +5°C to 150°C
S38781 120 V, 0.9 A, 110 W 1 Block Ambient +5°C to 150°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.
Catalog No. S38783 Supplier OHAUS™ Supplier No. 30392059
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.

Designed for applications that require repeatable results and superior temperature stability.

Dry block heaters require either one or two modular blocks, sold separately.

  • Ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures
  • Integral support rod holder with locking knob accepts optional external temperature probe kit
  • Accept separate interchangeable modular blocks (sold separately), accommodating various tube sizes from 0.2mL microtubes to 50mL centrifuge tubes
  • Each block has a thermometer well for measuring block temperature (blocks sold separately)
  • Efficient heater features close tube-and-block contact with a low-density design for exceptional temperature uniformity
  • Temperature range: ambient 5° to 120°C
  • With microprocessor control, LED display, audible alarm, temperature calibration mode

Applications:

Denaturing proteins, DNA applications, ELISA and other immunoassay studies

TRUSTED_SUSTAINABILITY

Specifications

Holds 1 Block
Temperature Range (Metric) Ambient +5° to 120°C
Type Dry Block Heater
For Use With (Application) Ideal for incubation and activation of cultures, Enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedure
Frame Material Steel
Heat Up Time 45 min.
Includes 92 in. Detachable, 3-wire cord and plug
Length (English) 12.4 in.
Length (Metric) 31.5 cm
Height (English) 3.5 in.
Height (Metric) 8.9 cm
Weight (English) 5.8 lb.
Weight (Metric) 2.6 kg
Electrical Requirements 120 V, 0.9 A, 110 W
Amperage 0.9 A
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.