Learn More
Thermco Products Inc Thermco NIST Compliant Block Heater/Water Bath Spirit Filled Laboratory Thermometer, 0 to 110C Range, 0.5C Divisions, 35mm Partial Immersion, 225mm Length
Catalog No. NC0673868
Encompass_Preferred
Precision spirit filled thermometer. The ACC6471S thermometer was designed to be used in block heater (dry baths) with it's 35mm immersion. Accurate to N.I.S.T. tolerances. Eligible for N.I.S.T. certification. Also used in shallow water baths
Catalog No. NC0673868 Supplier Thermco Products Inc Supplier No. ACC6471S
Precision spirit filled thermometer. The ACC6471S thermometer was designed to be used in block heater (dry baths) with it's 35mm immersion. Accurate to N.I.S.T. tolerances. Eligible for N.I.S.T. certification. Also used in shallow water baths