Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Dry Erase Board Cleaner
SCI_ED

Catalog No. S21600 Shop All Aldon Products
Change view
Click to view available options
Volume (English):
4 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
S21600 4 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S21600 Supplier Innovating Science™ Supplier No. IS5600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Safe cleaner removes shadows, ghosting and stains.

  • Just Spray and Wipe
  • Cleans and reconditions white boards
  • Safe cleaner removes shadows, ghosting and stains
  • 4 oz.

Specifications

Product Type Cleaner
Color-coded No
Form Liquid
Scent Unscented
Sterility Non-sterile
Appearance White
Container Type Spray bottle
Dimensions (L x W x H) 1.5 x 1.5 x 6.375 in.
For Use With (Application) Cleaning and conditioning white boards
For Use With (Equipment) White boards
Quantity 1 Ea.
Size 4 oz
Volume (English) 4 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.