Learn More
Stoelting™ Dry Glass Bead Sterilizer
SDP
Catalog No. 10000632 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-632 Neuroscience
1 options
Catalog No. 10-000-632 Supplier Stoelting™ Supplier No. 50287
Add to Cart
Add to Cart
Insert the cleaned surgical instrument for 2 to 5 seconds and remove. Within 30 seconds, the surgical instruments are cool enough to use.
Specifications
For Use With (Application) Neuroscience