Learn More
MESA LABS BOZEMAN MFG FACILITY CHEMICAL INDICATOR FOR DRY HEAT
SDP
Catalog No. NC9504938
Encompass
PROCHEM DRY HEAT INDICATOR LABELS – DRY HEAT TYPE 4 MULTIVARIABLE INDICATOR. Color change from green to dark brown/black within 10 minutes at 160°C. Distinguishes between processed and unprocessed items. Conforms to applicable portions of ISO 11140-1. One thousand (1,000) indicators/roll. 3 year expiry.
This item is not returnable. View return policy
Catalog No. NC9504938 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. 7705CIDHI
PROCHEM DRY HEAT INDICATOR LABELS – DRY HEAT TYPE 4 MULTIVARIABLE INDICATOR. Color change from green to dark brown/black within 10 minutes at 160°C. Distinguishes between processed and unprocessed items. Conforms to applicable portions of ISO 11140-1. One thousand (1,000) indicators/roll. 3 year expiry.