Learn More
SPS Medical DRY HEAT INDICATING
Catalog No. nc9921124
Encompass
Each chemical indicator (CI) strip provides distinct color change when exposed to the sterilization process. An internal CI should be used within each package or tray to be sterilized. Four inch strip can be written on and kept as a permanent record.
Catalog No. NC9921124 Supplier SPS Medical Supplier No. DIS100
Each chemical indicator (CI) strip provides distinct color change when exposed to the sterilization process. An internal CI should be used within each package or tray to be sterilized. Four inch strip can be written on and kept as a permanent record.