Learn More
SPS Medical DRY HEAT INDICATOR LABEL
Catalog No. nc0237998
Encompass
Labels are available in paper or universal stock. Universal labels can be used in steam, EO gas and hydrogen peroxide sterilization processes, allowing all packs to be labeled prior to sterilization. Multiple styles and colors available.
Catalog No. NC0237998 Supplier SPS Medical Supplier No. DIL1000
Labels are available in paper or universal stock. Universal labels can be used in steam, EO gas and hydrogen peroxide sterilization processes, allowing all packs to be labeled prior to sterilization. Multiple styles and colors available.