Learn More
BURKLE INC SnowPack 50 dry ice maker, tablet Ø x h 1.97x1.18 in. (50x30 mm)
Catalog No. 5000025648
Encompass_Preferred
SnowPack enables you to obtain a -79 °C (-110.2 °F) dry ice tablet weighing 50 or 250 g (1.76 or 8.82 oz.) within a few moments, without complicated technology and without electricity! You can therefore obtain low-cost cooling to very low temperatures at any time you like.
Dry ice is easy to handle, odorless and flavorless. The SnowPack dry ice machine can be installed on any commercial CO2-bottle with a standpipe (without pressure regulator) (thread W 21.80 x 1/14" according to DIN 477-1 No. 6). These often exist in many production facilities and laboratories anyway. If this is not the case, a bottle can be rented for a small price from a local specialist gas distributor.
Catalog No. 50-000-25648 Supplier BURKLE INC Supplier No. 98050030
SnowPack enables you to obtain a -79 °C (-110.2 °F) dry ice tablet weighing 50 or 250 g (1.76 or 8.82 oz.) within a few moments, without complicated technology and without electricity! You can therefore obtain low-cost cooling to very low temperatures at any time you like.
Dry ice is easy to handle, odorless and flavorless. The SnowPack dry ice machine can be installed on any commercial CO2-bottle with a standpipe (without pressure regulator) (thread W 21.80 x 1/14" according to DIN 477-1 No. 6). These often exist in many production facilities and laboratories anyway. If this is not the case, a bottle can be rented for a small price from a local specialist gas distributor.