Learn More
Labelmaster Inc Category A Biological Substance Dry Ice Packaging
SDP
Catalog No. NC9941684
Encompass_Preferred
This Dry Ice Packaging may be used for shipping infectious materials that must be kept cold during transport. The infectious substance shipper holds up to five vials of infectious materials with a total gross weight of 94g.
Catalog No. NC9941684 Supplier Labelmaster Inc Supplier No. HMS69110
This Dry Ice Packaging may be used for shipping infectious materials that must be kept cold during transport. The infectious substance shipper holds up to five vials of infectious materials with a total gross weight of 94g.