Learn More
Cole-Parmer Dry Ice Storage Chest; 3.75 cu ft, 200 lbs Capacity, 19-1/8" x 25-1/4" x 28-1/8"
Catalog No. NC2010679 Shop All Cole-Parmer Products
Encompass_Preferred
Double-wall polyethylene construction filled with urethane foam insulatio. Perfect for short or long-term storage Ideal for dispensing dry ice in a laboratory environment Rounded interior corners for easy clean-up. Wall thickness of 2". Large-Capacity dry ice storage chests feature a lightweight construction with special formulated insulation for lowest sublimation rates. Chests contain extra-thick foam plastic insulation core. It takes 6 days for 85 lb of dry ice to sublimate to 5 lb in model 03721-02; 7 days for 150 lb in model 03721-00; and 12 days for 200 lb in model 03720-00.
Catalog No. NC2010679 Supplier Cole-Parmer Supplier No. 0372000
Double-wall polyethylene construction filled with urethane foam insulatio. Perfect for short or long-term storage Ideal for dispensing dry ice in a laboratory environment Rounded interior corners for easy clean-up. Wall thickness of 2". Large-Capacity dry ice storage chests feature a lightweight construction with special formulated insulation for lowest sublimation rates. Chests contain extra-thick foam plastic insulation core. It takes 6 days for 85 lb of dry ice to sublimate to 5 lb in model 03721-02; 7 days for 150 lb in model 03721-00; and 12 days for 200 lb in model 03720-00.