Learn More
Cole-Parmer ThermoSafe 450 Dry Ice Storage Chest; 1.6 cu ft, 85 lb Pellet Capacity
Catalog No. NC1488074 Shop All Cole-Parmer Products
Encompass_Preferred
Large-capacity dry ice storage chests feature a lightweight construction with special formulated insulation for lowest sublimation rates. Chests contain extra-thick foam plastic insulation core. It takes 6 days for 85 pounds of dry ice to sublimate to 5 pounds. The chest has handles for easy carrying. Order dollies separately for improved mobility.
Catalog No. NC1488074 Supplier Cole-Parmer Supplier No. 0372102
Large-capacity dry ice storage chests feature a lightweight construction with special formulated insulation for lowest sublimation rates. Chests contain extra-thick foam plastic insulation core. It takes 6 days for 85 pounds of dry ice to sublimate to 5 pounds. The chest has handles for easy carrying. Order dollies separately for improved mobility.