Learn More
Saf T Pak Inc STP-714 - Disposable 2-Part Secondary Pressure Receptacle & Leakproof Bag, Medium
SDP
Catalog No. NC9447910 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
Disposable 2-Part Secondary Pressure Receptacle & Leakproof Bag, Designed to allow direct contact with Dry Ice, Inner Leakproof Polybag ID 8.5 x 10.5in. Outer Tyvek Envelope dimensions 7.5 x 9.5in. 50/Case
Catalog No. NC9447910 Supplier Saf T Pak Inc Supplier No. STP714
Disposable 2-Part Secondary Pressure Receptacle & Leakproof Bag, Designed to allow direct contact with Dry Ice, Inner Leakproof Polybag ID 8.5 x 10.5in. Outer Tyvek Envelope dimensions 7.5 x 9.5in. 50/Case