Learn More
Cole-Parmer Dry-Keeper H420580001 Desiccator Cabinets, Horizontal; Non-Electric
Catalog No. NC1501596 Shop All Cole-Parmer Products
Encompass
Non-Electric Desiccator Cabinets are available in horizontal, vertical, or stacking models. Horizontal and vertical models are constructed of clear acrylic. The smoke-tinted polystyrene stacking desiccator can be stacked up to five high to save space.
Catalog No. NC1501596 Supplier Cole-Parmer Supplier No. 0890950
Non-Electric Desiccator Cabinets are available in horizontal, vertical, or stacking models. Horizontal and vertical models are constructed of clear acrylic. The smoke-tinted polystyrene stacking desiccator can be stacked up to five high to save space.