Learn More
Electron Microscopy Sciences Dry Keeper Plus™ Desiccator Cabinet; Non-Vacuum
SDP
Catalog No. 501899510
Encompass_Preferred
Dry Keeper Plus Dessicator cabinet features a user-friendly electronic desiccant control, one-touch door latch with silicone rubber door gasket, and molded cabinet made from ABS plastic. Efficient dehumidification is monitored by a hygrometer dial in the door. The dehumidifying auto-drying unit has an air circulating system which helps drop relative humidity sharply through its cycling of absorption and regeneration of the enclosed permanent desiccant. It can bring the relative humidity down from 60% to 10% in 6 hours of continuous operation in an unloaded cabinet.
The unit comes with three clear, removable shelves with vent holes which provide evenly spaced storage area. Comes with a 2-prong 6 ft (2 meter) cord.
Catalog No. 50-189-9510 Supplier Electron Microscopy Sciences Supplier No. 7121800
Dry Keeper Plus Dessicator cabinet features a user-friendly electronic desiccant control, one-touch door latch with silicone rubber door gasket, and molded cabinet made from ABS plastic. Efficient dehumidification is monitored by a hygrometer dial in the door. The dehumidifying auto-drying unit has an air circulating system which helps drop relative humidity sharply through its cycling of absorption and regeneration of the enclosed permanent desiccant. It can bring the relative humidity down from 60% to 10% in 6 hours of continuous operation in an unloaded cabinet.
The unit comes with three clear, removable shelves with vent holes which provide evenly spaced storage area. Comes with a 2-prong 6 ft (2 meter) cord.