Learn More
Biotage Dry Load 70g Cartidge - 20 per Pack
SDP
Catalog No. NC0824852
Encompass
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. With maximum flexibility, users place the needed amount of adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20 with 20 frits. These are empty vessels and can accomodate up to approximately 70 grams of most sorbants, such as silica gel.Compatible with earlier versions of Biotage DLV part number DLV-070, These are not compatible with current version DLV plunger DLV-050Note: These vessels come equipped with luer fittings and do not have twist locking capability
Catalog No. NC0824852 Supplier Biotage Supplier No. DLV075
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. With maximum flexibility, users place the needed amount of adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20 with 20 frits. These are empty vessels and can accomodate up to approximately 70 grams of most sorbants, such as silica gel.Compatible with earlier versions of Biotage DLV part number DLV-070, These are not compatible with current version DLV plunger DLV-050Note: These vessels come equipped with luer fittings and do not have twist locking capability