Learn More
StatLab Dry Mats Formalin Neutralizing Pads
SDP
Catalog No. 22000003
Change view
Click to view available options
Material:
Cellulose, Polyester
Catalog No. Material
22-000-003 Cellulose, Polyester
1 options
Catalog No. 22-000-003 Supplier StatLab Supplier No. 172854
Add to Cart
Edge
Add to Cart
A unique solution for aggressive spill response and formalin vapor control.
  • Highly absorbent and dry 12 x 13 in. sheets contain a proprietary formaldehyde-specific neutralizer
  • Excellent for grossing stations, countertops, storage cabinets, and any area where formaldehyde spills might occur.
Specifications
Material Cellulose, Polyester
Type Absorbent Pad
Disposable Limited-use
Weight Capacity Heavyweight