Learn More
AG Scientific Inc Dry Powder Milk 1 KG
Catalog No. 502268287
Encompass_Preferred
Nonfat dry milk for use as a blocking agent during Western Blotting to block non-specific antibody binding and reduce background signal.
Catalog No. 50-226-8287 Supplier AG Scientific Inc Supplier No. D25871KG
Nonfat dry milk for use as a blocking agent during Western Blotting to block non-specific antibody binding and reduce background signal.