Learn More
Glas Col LLC Rotator
SDP
Supplier:  Glas Col LLC 099A RD9912
Encompass_Preferred
Dry powder rotator. RPM 2-80. 120V. For bottles 7" to 10" in height and up to 7" in diameter. Ten pound maximum weight capacity. Variable speed control. 1.5A, 50/60 Hz.
Catalog No. NC9842224