Learn More
Epredia™ Dry Powder Stains
Catalog No. 22110868 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Eosin
Wright Stain
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-110-868 Wright Stain
22-110-837 Eosin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-110-868 Supplier Epredia™ Supplier No. 90049
Add to Cart
Add to Cart
This item is not returnable. View return policy
Stain specimens with Eosin-Y or Wright compounds with Epredia™ Dry Powder Stains, certified by the Biological Stain Commission.
Order Info
Quantity: 100 g
Specifications
Quantity 100 g
Product Type Wright Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov