Learn More
Mopec Dry Shield Faraday Tote
SDP
Catalog No. 22446161 Shop All Mopec Products
Change view
Click to view available options
Material:
Fabric
Catalog No. Material
22-446-161 Fabric
1 options
Catalog No. 22-446-161 Supplier Mopec Supplier No. FS148
Add to Cart
Add to Cart
Waterproof bag that blocks all wireless signals to keep devices offline and secure
  • Features a Roll-top dry bag closure with side buckles, an unshielded waterproof clear front zipper pocket, and a Load-bearing, adjustable, detachable shoulder strap.
  • Folds flat for convenient transport and storage, shields laptops, tablets, cell phones, radios, and accessories, and protects against rain, snow, sand, dirt, and contaminants.
  • Protects electronics from tracking, hacking, and remote access
  • Provides privacy, security, and safe device transport.
  • Constructed with two layers of High-shielding TitanRF Faraday Fabric and 500D PVC.
  • Internal usable dimensions when closed measure 17 x 9 in.
Specifications
Product Type Tote
Color Black
Material Fabric
Sterility Non-sterile