Learn More
Mitegen Dry Shipper Air Dryer for Efficient Moisture Removal
SDP
Catalog No. 5000495016
Encompass_Preferred
The Dry Shipper Air Dryer is an essential accessory for maintaining the performance and longevity of dry shippers, such as the CX100. Dry shippers can accumulate ice and moisture, which reduces their liquid nitrogen (LN2) holding capacity and compromises their insulating properties. While warming a dry shipper melts this ice, the Dry Shipper Air Dryer is specifically designed to efficiently remove any residual moisture, ensuring the dry shipper is thoroughly dry before recharging and subsequent use. For optimal results, it should be used in conjunction with the MiTeGen Dry Shipper Warming and Drying Racks, see SKUs ML-DR-CX100-2 and ML-DR-CX100-4. This tool utilizes two small centrifugal blowers to provide a gentle, integrated heated airflow that targets and removes residual moisture from the adsorbent material without causing any damage. It is an essential tool for maximizing holding times and protecting the integrity of your samples.
Catalog No. 50-004-95016 Supplier Mitegen Supplier No. MCXAD01
The Dry Shipper Air Dryer is an essential accessory for maintaining the performance and longevity of dry shippers, such as the CX100. Dry shippers can accumulate ice and moisture, which reduces their liquid nitrogen (LN2) holding capacity and compromises their insulating properties. While warming a dry shipper melts this ice, the Dry Shipper Air Dryer is specifically designed to efficiently remove any residual moisture, ensuring the dry shipper is thoroughly dry before recharging and subsequent use. For optimal results, it should be used in conjunction with the MiTeGen Dry Shipper Warming and Drying Racks, see SKUs ML-DR-CX100-2 and ML-DR-CX100-4. This tool utilizes two small centrifugal blowers to provide a gentle, integrated heated airflow that targets and removes residual moisture from the adsorbent material without causing any damage. It is an essential tool for maximizing holding times and protecting the integrity of your samples.