Learn More
Mitegen Dry Shipper Warming and Drying Rack, 4 Dry Shipper Capacity
SDP
Catalog No. 5000494987
Encompass_Preferred
The Dry Shipper Warming and Drying Rack is an essential tool for maintaining the efficiency and longevity of your dry shippers, such as the CX100. This rack holds 4 dry shippers. Over time, ice can accumulate within dry shippers, and when it melts, the resulting water can become trapped in the adsorbent material. This trapped water reduces the shipper's holding time and can compromise the integrity of your samples. This rack addresses the issue by providing a stable and convenient platform to warm and dry your dry shippers, ensuring optimal performance. It securely holds the shipper vertically and upside-down to maximize the rate of natural convective warming and to ensure water drains out effectively. The rack is available in two-unit and four-unit versions, is constructed from durable stainless steel, and is designed with mobility in mind, featuring lockable wheels. The base of the rack also includes a convenient lower storage area to keep all components organized.
Catalog No. 50-004-94987 Supplier Mitegen Supplier No. MDRCX1004
Add to Cart
Add to Cart
The Dry Shipper Warming and Drying Rack is an essential tool for maintaining the efficiency and longevity of your dry shippers, such as the CX100. This rack holds 4 dry shippers. Over time, ice can accumulate within dry shippers, and when it melts, the resulting water can become trapped in the adsorbent material. This trapped water reduces the shipper's holding time and can compromise the integrity of your samples. This rack addresses the issue by providing a stable and convenient platform to warm and dry your dry shippers, ensuring optimal performance. It securely holds the shipper vertically and upside-down to maximize the rate of natural convective warming and to ensure water drains out effectively. The rack is available in two-unit and four-unit versions, is constructed from durable stainless steel, and is designed with mobility in mind, featuring lockable wheels. The base of the rack also includes a convenient lower storage area to keep all components organized.