Learn More
Fritsch Milling And Sizing Inc Dry Sieve Pan 8" Stl. Steel
SDP
Catalog No. 501534169
Encompass
For dry sieving with test sieves 8" dia.
The sieve pan made of stainless steel completes the sieve stack.
Catalog No. 50-153-4169 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.1020.03
For dry sieving with test sieves 8" dia.
The sieve pan made of stainless steel completes the sieve stack.