Learn More
Andwin Scientific Yamato 159L Dry Sterilizer, Natural Convection, 115V/220V
SDP
Catalog No. 501898074
Encompass_Preferred
Programmable 159L dry heat sterilizer with improved temperature controller and enhanced safety features and independent overheat prevention device. Digital setting by main menu and ▲▼ keys for fixed temperature, program, quick auto stop, auto stop and auto start operations. Standard equipped with calibration off-set, lock function, power failure recovery mode. Programmable with maximum 30 steps, 6 patterns, repeat operation. Operating temp. range ambient +5~260°C
Catalog No. 50-189-8074 Supplier Andwin Scientific Supplier No. SI601
Programmable 159L dry heat sterilizer with improved temperature controller and enhanced safety features and independent overheat prevention device. Digital setting by main menu and ▲▼ keys for fixed temperature, program, quick auto stop, auto stop and auto start operations. Standard equipped with calibration off-set, lock function, power failure recovery mode. Programmable with maximum 30 steps, 6 patterns, repeat operation. Operating temp. range ambient +5~260°C