Learn More
Yamato Scientific America, Inc. Yamato 99L Dry Sterilizer, Natural Convection, 115V/220V
Catalog No. 501898075 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Programmable natural convection 99L dry heat sterilizer with independent overheat prevention device. High precision controller with improved display visibility and operability. Standard equipped with calibration off-set, lock function, power recovery mode, power on and operation time accumulation, calendar time, accumulation power consumption monitoring, total CO₂ emission, and heat output, save and access operator setting information. Maximum 99 steps, 99 patterns, repeat operation. Easy sample data collection with cable port. Temp setting range ambient +5~260°C.
Catalog No. 50-189-8075 Supplier Yamato Scientific America, Inc. Supplier No. SK401115V / SK401220V
Programmable natural convection 99L dry heat sterilizer with independent overheat prevention device. High precision controller with improved display visibility and operability. Standard equipped with calibration off-set, lock function, power recovery mode, power on and operation time accumulation, calendar time, accumulation power consumption monitoring, total CO₂ emission, and heat output, save and access operator setting information. Maximum 99 steps, 99 patterns, repeat operation. Easy sample data collection with cable port. Temp setting range ambient +5~260°C.