Learn More
Cole-Parmer Fluke Calibration 9140-A Metrology Dry-Well Calibrator
Catalog No. 50993428 Shop All Cole-Parmer Products
Encompass
These new units beat every other comparable dry-well in the industry in performance, size, weight, convenience, ease of calibration, software and price. In addition, the heating and cooling rate of each of these dry-wells is adjustable from the front panel, thermal switches can be checked by these units for actuation testing and multiple-hole inserts are available for a variety of probe sizes. Choose one of three removable inserts sized from 1/16" to 1/2" diameter. Insert A handles a full range of probe sizes with a single well of each size. Insert B features two wells each of 3/8", 1/4" and 3/16" in diameter for doing comparison calibrations, and insert C has six 1/4" diameter wells for multiple probe calibrations.
FEATURES
  • Accuracy to ± 0.5 °C
  • RS232 and Windows software included
  • Easy to recalibrate
  • Less than eight pounds
Catalog No. 50-993-428 Supplier Cole-Parmer Supplier No. 1610170
These new units beat every other comparable dry-well in the industry in performance, size, weight, convenience, ease of calibration, software and price. In addition, the heating and cooling rate of each of these dry-wells is adjustable from the front panel, thermal switches can be checked by these units for actuation testing and multiple-hole inserts are available for a variety of probe sizes. Choose one of three removable inserts sized from 1/16" to 1/2" diameter. Insert A handles a full range of probe sizes with a single well of each size. Insert B features two wells each of 3/8", 1/4" and 3/16" in diameter for doing comparison calibrations, and insert C has six 1/4" diameter wells for multiple probe calibrations.
FEATURES
  • Accuracy to ± 0.5 °C
  • RS232 and Windows software included
  • Easy to recalibrate
  • Less than eight pounds