Learn More
ORS Nasco DRYBOX GLV AMBI 15MIL 9.75DRY
Catalog No. 502551652
Encompass_Preferred
Honeywell part# 8Y1532A/9Q. Honeywell CSM Glove Box Gloves 8 in. Port Circumference Hand: 9.75 in. 25 cm Chlorosulfonated Polyethylene (CSM) Hand: Ambidextrous Thickness: 15 mil 0.38 mm White Latex Free: Yes Port Size: 20.3 cm 8 in. Size: 9.75 Certifications/Compliance: EN 388 EN 374 Exterior Finish: Smooth
Catalog No. 50-255-1652 Supplier ORS Nasco Supplier No. 0688Y1532A/9Q
Honeywell part# 8Y1532A/9Q. Honeywell CSM Glove Box Gloves 8 in. Port Circumference Hand: 9.75 in. 25 cm Chlorosulfonated Polyethylene (CSM) Hand: Ambidextrous Thickness: 15 mil 0.38 mm White Latex Free: Yes Port Size: 20.3 cm 8 in. Size: 9.75 Certifications/Compliance: EN 388 EN 374 Exterior Finish: Smooth