Learn More
Research Products International Corp DryChill Blocks 40x1.5-2.0ml
SDP
Catalog No. 502388418
Encompass
The DryChill cooling blocks are designed to store samples safely on the lab-bench without sample degradation due to temperature increase or fluctuation. Unlike traditional ice buckets, these blocks do not require any ice and are uniquely designed to minimize condensation, resulting in convenient, mess-free cooling. Each DryChill Cooling Block utilizes a proprietary gel refrigerant, designed to maintain cold (but not freezing) temperatures for extended time periods. Once the block temperature begins to warm, the visual indicator alerts the user by changing from purple to pink color.
Ice-free design reduces condensation
Maintains 4°C up to 200 minutes
Visual temperature indicator
Catalog No. 50-238-8418 Supplier Research Products International Corp Supplier No. 400700
The DryChill cooling blocks are designed to store samples safely on the lab-bench without sample degradation due to temperature increase or fluctuation. Unlike traditional ice buckets, these blocks do not require any ice and are uniquely designed to minimize condensation, resulting in convenient, mess-free cooling. Each DryChill Cooling Block utilizes a proprietary gel refrigerant, designed to maintain cold (but not freezing) temperatures for extended time periods. Once the block temperature begins to warm, the visual indicator alerts the user by changing from purple to pink color.
Ice-free design reduces condensation
Maintains 4°C up to 200 minutes
Visual temperature indicator