Learn More
Stellar Scientific Benchmark Scientific DryChill Cooling Block, Iceless, 40 x 1.5/2.0 mL Tubes, 4°C for 210 min
SDP
Catalog No. NC3973317
Encompass_Preferred
The DryChill cooling blocks are designed to store samples safely on the lab-bench without sample degradation due to temperature increase or fluctuation. Unlike traditional ice buckets, these blocks do not require any ice and are uniquely designed to minimize condensation, resulting in a convenient, mess-free cooling. Each DryChill Cooling Block utilizes a proprietary gel refrigerant, designed to maintain cold (but not freezing) temperatures for extended time periods. Once the block temperature begins to warm, the visual indicator alerts the user by changing from purple to pink. Four blocks are available for accepting a wide variety of commonly used research tubes and plates.
  • Ice-free, stable cooling for sample tubes
  • Unique design reduces condensation
  • Maintains 4°C for up to +200 minutes
  • Four sizes for various tubes up to 50 mL
  • Visual temperature indicator
Catalog No. NC3973317 Supplier Stellar Scientific Supplier No. BSDC40
The DryChill cooling blocks are designed to store samples safely on the lab-bench without sample degradation due to temperature increase or fluctuation. Unlike traditional ice buckets, these blocks do not require any ice and are uniquely designed to minimize condensation, resulting in a convenient, mess-free cooling. Each DryChill Cooling Block utilizes a proprietary gel refrigerant, designed to maintain cold (but not freezing) temperatures for extended time periods. Once the block temperature begins to warm, the visual indicator alerts the user by changing from purple to pink. Four blocks are available for accepting a wide variety of commonly used research tubes and plates.
  • Ice-free, stable cooling for sample tubes
  • Unique design reduces condensation
  • Maintains 4°C for up to +200 minutes
  • Four sizes for various tubes up to 50 mL
  • Visual temperature indicator