Learn More
MSC Makita Dry Dust Extractor/Vacuum Kit, 36V (18V X2) LXT/Corded, 2.1 Gallon, HEPA Filter
Catalog No. 502837319
Encompass
The 36V (18V X2) LXT/Corded 2.1 Gallon HEPA Filter Dry Dust Extractor/Vacuum Kit offers versatile corded and cordless operation. It features a powerful motor and a three-stage HEPA filtration system capturing 99.97% of particulates 0.3 microns and larger, designed for portability and durability.
  • Corded or cordless operation.
  • Up to 65 minutes run time on low setting with two 18V 5.0Ah batteries.
  • Three-stage HEPA filtration captures 99.97% of particulates 0.3 microns and larger.
  • Motor delivers 127 CFM and 96" water lift (corded), or 74 CFM and 36" water lift (battery).
  • Lightweight at 21.8 lbs with batteries.
  • Star protection computer controls™ protect against overloading and overheating.
  • Sound-absorbing motor chamber for quiet operation.
Catalog No. 50-283-7319 Supplier MSC Supplier No. 42247742
The 36V (18V X2) LXT/Corded 2.1 Gallon HEPA Filter Dry Dust Extractor/Vacuum Kit offers versatile corded and cordless operation. It features a powerful motor and a three-stage HEPA filtration system capturing 99.97% of particulates 0.3 microns and larger, designed for portability and durability.
  • Corded or cordless operation.
  • Up to 65 minutes run time on low setting with two 18V 5.0Ah batteries.
  • Three-stage HEPA filtration captures 99.97% of particulates 0.3 microns and larger.
  • Motor delivers 127 CFM and 96" water lift (corded), or 74 CFM and 36" water lift (battery).
  • Lightweight at 21.8 lbs with batteries.
  • Star protection computer controls™ protect against overloading and overheating.
  • Sound-absorbing motor chamber for quiet operation.