Learn More
Chemglass Life Sciences Drying Apparatus, Abderhalden, Complete, 740mm Height x 350mm Width
SDP
Catalog No. CG128901
Encompass_Preferred
Drying Apparatus, Abderhalden, Complete, 740mm Height x 350mm Width. Abderhalden style for drying samples at a temperature as a function of the solvent boiling point. The desiccant flask has a 34/45 outer joint for connection to the drying chamber. All other joints are 24/40. Complete apparatus consists of a desiccant flask, drying chamber, round bottom flask and Allihn condenser.
Catalog No. CG128901 Supplier Chemglass Life Sciences Supplier No. CG128901
Drying Apparatus, Abderhalden, Complete, 740mm Height x 350mm Width. Abderhalden style for drying samples at a temperature as a function of the solvent boiling point. The desiccant flask has a 34/45 outer joint for connection to the drying chamber. All other joints are 24/40. Complete apparatus consists of a desiccant flask, drying chamber, round bottom flask and Allihn condenser.