Learn More
Anton Paar DRYING CARTRIDGE ABBEMAT PL PPL 3X00
Catalog No. NC2736775
Encompass_Preferred
Compatible with: Abbemat Performance 300, Abbemat Performance 500, Abbemat Performance Plus 350, Abbemat Performance Plus 550, Abbemat 3000, Abbemat 3100, Abbemat 3200, Brix refractometer: Abbemat Juice Station
Catalog No. NC2736775 Supplier Anton Paar Supplier No. 107786
Compatible with: Abbemat Performance 300, Abbemat Performance 500, Abbemat Performance Plus 350, Abbemat Performance Plus 550, Abbemat 3000, Abbemat 3100, Abbemat 3200, Brix refractometer: Abbemat Juice Station