Learn More
Chemglass Life Sciences Drying Chamber, #50 O-Ring Joint, Lower Vessel 50mm ID X 100mm Height, Airfree, Schlenk, #75 Clamp Size, #229 O-Ring Size
SDP
Catalog No. NC1420889
Encompass_Preferred
Chamber for use in drying small quantities of material in dram vials. Can also be used in the transfer of materials from glovebox to glovebox or to the vacuum line. Lower vessel has a flat bottom permitting it to be stood upright on the bench or in a glovebox. Top adapter has a 0-4mm Chem-Vac Chem-Cap valve. Complete item is supplied with the corresponding CG-150 S.S. pinch clamp and one Viton o-ring.
Catalog No. NC1420889 Supplier Chemglass Life Sciences Supplier No. AF055602
Chamber for use in drying small quantities of material in dram vials. Can also be used in the transfer of materials from glovebox to glovebox or to the vacuum line. Lower vessel has a flat bottom permitting it to be stood upright on the bench or in a glovebox. Top adapter has a 0-4mm Chem-Vac Chem-Cap valve. Complete item is supplied with the corresponding CG-150 S.S. pinch clamp and one Viton o-ring.