Learn More
Chemglass Life Sciences Drying Chamber, #40 O-Ring Joint, Lower Vessel 40mm ID X 100mm Height, Airfree, Schlenk, #65 Clamp Size, #226 O-Ring Size
SDP
Catalog No. AF055601
Encompass_Preferred
Drying Chamber, #40 O-Ring Joint, Lower Vessel 40mm ID X 100mm Height, Airfree, Schlenk, #65 Clamp Size, #226 O-Ring Size. Chamber for use in drying small quantities of material in dram vials. Can also be used in the transfer of materials from glovebox to glovebox or to the vacuum line. Lower vessel has a flat bottom permitting it to be stood upright on the bench or in a glovebox. Top adapter has a 0-4mm Chem-Vac™ Chem-Cap valve. Complete item is supplied with the corresponding CG-150 Stainless Steel pinch clamp and one Viton o-ring.
Catalog No. AF055601 Supplier Chemglass Life Sciences Supplier No. AF055601
Drying Chamber, #40 O-Ring Joint, Lower Vessel 40mm ID X 100mm Height, Airfree, Schlenk, #65 Clamp Size, #226 O-Ring Size. Chamber for use in drying small quantities of material in dram vials. Can also be used in the transfer of materials from glovebox to glovebox or to the vacuum line. Lower vessel has a flat bottom permitting it to be stood upright on the bench or in a glovebox. Top adapter has a 0-4mm Chem-Vac™ Chem-Cap valve. Complete item is supplied with the corresponding CG-150 Stainless Steel pinch clamp and one Viton o-ring.