Learn More
Chemglass Life Sciences Drying Chamber Only, 24/40
SDP
Catalog No. CG129006
Encompass_Preferred
Drying chamber only, 24/40, for the CG129005 Abderhalden drying apparatus. The drying chamber has an I.D. of 2” and is 8” long.
Catalog No. CG129006 Supplier Chemglass Life Sciences Supplier No. CG129006
Drying chamber only, 24/40, for the CG129005 Abderhalden drying apparatus. The drying chamber has an I.D. of 2” and is 8” long.