Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Drying Column with Reservoir

Catalog No. 111016088 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Sample Capacity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Capacity
11-101-6088 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6088 Supplier MilliporeSigma™ Supelco™ Supplier No. 64785U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meets US EPA specifications for drying solvent extracts in procedures for semivolatile organic compounds. Reservoir holds approximately 60 mL of extract.

  • Meets US EPA specifications for drying solvent extracts in procedures for semivolatile organic compounds
  • Reservoir holds approximately 60 mL of extract, the entire product from one separatory funnel extraction
  • Holds a 19 x 100 mm bed of sodium sulfate
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance US EPA Specifications for Drying Solvent Extracts
Description Holds a 19 x 100 mm Bed of Sodium Sulfate
Type Drying Column with Reservoir
Sample Capacity 60 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.