Learn More
Chemglass Life Sciences Chemglass Heavy Wall Drying Dish, 457 mm x 305 mm x 72 mm
SDP
Catalog No. NC3355230
Encompass_Preferred
The Chemglass Heavy Wall Drying Dish is a Pyrex glass tray designed for laboratory drying, staining, and developing applications. It withstands hot-air and steam sterilization and is suitable for gel work and paper chromatography. Do not place over direct heat; intended for oven use only.
  • Heavy-wall construction for increased durability.
  • Dimensions: 457 mm x 305 mm x 72 mm.
  • Heat resistant; compatible with hot-air and steam sterilization.
  • Suitable for gel work, staining, and paper chromatography.
  • Oven use only; do not place over direct heat.
  • Reusable laboratory drying tray for general lab applications.
Catalog No. NC3355230 Supplier Chemglass Life Sciences Supplier No. CG828012
The Chemglass Heavy Wall Drying Dish is a Pyrex glass tray designed for laboratory drying, staining, and developing applications. It withstands hot-air and steam sterilization and is suitable for gel work and paper chromatography. Do not place over direct heat; intended for oven use only.
  • Heavy-wall construction for increased durability.
  • Dimensions: 457 mm x 305 mm x 72 mm.
  • Heat resistant; compatible with hot-air and steam sterilization.
  • Suitable for gel work, staining, and paper chromatography.
  • Oven use only; do not place over direct heat.
  • Reusable laboratory drying tray for general lab applications.